Fisher Scientific™ Precision Specific Gravity Hydrometers, 340mm Long
Requires 275mL liquid volume for submersion
Manufacturer:  Fisher Scientific™ K11555CC
 View more versions of this product
Catalog No. 11-555CC
Specifications
Description
Hydrometer, Specific gravity; Fisherbrand; Precision; Universal range; Range: 0.760 to 0.830; Length: 330mm; With divisions of 0.0005; Use Cylinder 08-530K; W/3-pt certification
  • Use Cylinder 08-530K, available separately
  • Permanently marked with individual serial numbers
  • Accuracy meets or exceeds ASTM E 100 specifications
  • Calibrated according to ASTM E 126 specifications
  • With divisions of 0.001
  • Use Cylinder 08-530E, available separately
Description & Specifications
Specifications
0.0005
350mL Borosilicate Glass cylinder
330mm
0.76 to 0.83
3-point Certification, NIST, ASTM E100, ASTM E126