Please login to your online account to display your discounted pricing

Fisher Scientific™ Precision Specific Gravity Hydrometers, 340mm Long


Requires 275mL liquid volume for submersion

Manufacturer: fisher scientific™  11555Q

 View more versions of this product

Catalog No. 11-555Q

  • / Each
 

Description & Specifications

Specifications

Increments 0.0005
For Use With (Equipment) 350mL Borosilicate Glass cylinder
Length (Metric) 330mm
Certifications/Compliance NIST, ASTM E100, ASTM E126
Specific Gravity Range 1.48 to 1.55
Accessories

Accessories for Catalog Number: 11555Q

Item Mfr. No. Item Description Price Per Unit Quantity  
11-582-1
Thermo Scientific™ Nalgene™ PMP Hydrometer Jar
thermo scientific™
N62300500
Protect against breakage. This PMP jar is lighter than glass and offers superior chemical resistance and a broad base, making it a stable addition to the lab or field. Each for $80.17
Case of 6 Each for $481.02