Please login to your online account to display your discounted pricing

Fisher Scientific™ Precision Specific Gravity Hydrometers, 340mm Long


Requires 275mL liquid volume for submersion

Manufacturer: fisher scientific™  11555E

 View more versions of this product

Catalog No. 11-555E

  • / Each
 

Description & Specifications

Specifications

Increments 0.0005
Certifications/Compliance NIST, ASTM E100, ASTM E126
Length (Metric) 340mm
For Use With (Equipment) 350mL Borosilicate Glass cylinder
Specific Gravity Range 0.88 to 0.95

  • Use Cylinder 08-530K, available separately
  • Permanently marked with individual serial numbers
  • Accuracy meets or exceeds ASTM E 100 specifications
  • Calibrated according to ASTM E 126 specifications
  • With divisions of 0.001
  • Use Cylinder 08-530E, available separately

Accessories

Accessories for Catalog Number: 11555E

Item Mfr. No. Item Description Price Per Unit Quantity  
11-582-1
Thermo Scientific™ Nalgene™ PMP Hydrometer Jar
thermo scientific™
N62300500
Protect against breakage. This PMP jar is lighter than glass and offers superior chemical resistance and a broad base, making it a stable addition to the lab or field. Each for $80.17
Case of 6 Each for $481.02