Fisherbrand™ Precision Specific Gravity 150 - 185mm Long Hydrometers
Requires 140mL liquid volume for submersion.
Description
Requires 140mL liquid volume for submersion.
Used where a high degree of accuracy is required in hydrometer readings
- Permanently marked with individual serial numbers
- Accuracy meets or exceeds ASTM E 100 specifications
- Calibrated according to ASTM E 126 specifications
- With divisions of 0.001
- Use Cylinder 08-530E, available separately
Certifications
A certificate is provided to indicate traceability to standards provided by NIST™.
Specifications
0.001 | |
165 mm | |
Measure the True Specific Gravity of Liquids that are Lighter than Water | |
140 mL |
3-point Certification, NIST, ASTM E100, ASTM E126 | |
Soda Lime Glass | |
0.940 to 1.010 |