Please login to your online account to display your discounted pricing
Fisherbrand™ Precision Specific Gravity Hydrometers, 150mm Long
Requires 140mL liquid volume for submersion
Manufacturer: fisherbrand™  11556DC
 View more versions of this product
Catalog No. 11556DC
  • / Each
Description & Specifications
Specifications
Increments 0.001
Certifications/Compliance 3-point Certification, NIST, ASTM E100, ASTM E126
Length (Metric) 170mm
Specific Gravity Range 0.88 to 0.95