Fisherbrand™ Precision Specific Gravity Hydrometers, 150mm Long
Requires 140mL liquid volume for submersion
Manufacturer:  Fisherbrand™ 11556BC
 View more versions of this product
Catalog No. 11-556BC
Specifications Description & Specifications
Specifications
0.001
3-point Certification, NIST, ASTM E100, ASTM E126
160mm
0.76 to 0.83