Fisherbrand™ Precision Specific Gravity Hydrometers, 150mm Long
Requires 140mL liquid volume for submersion
Manufacturer:  Fisherbrand™ 11556R
 View more versions of this product
Catalog No. 11-556R
Specifications Description & Specifications
Specifications
0.001
NIST, ASTM E100, ASTM E126
165mm
1.6 to 1.67