Please login to your online account to display your discounted pricing

Fisherbrand™ Precision Specific Gravity Hydrometers, 150mm Long


Requires 140mL liquid volume for submersion

Manufacturer: fisherbrand™  11556F

 View more versions of this product

Catalog No. 11-556F

  • / Each
 

Description & Specifications

Specifications

Increments 0.001
Certifications/Compliance NIST, ASTM E100, ASTM E126
Length (Metric) 170mm
Specific Gravity Range 1 to 1.07
Accessories

Accessories for Catalog Number: 11556F

Item Mfr. No. Item Description Price Per Unit Quantity  
11-582-1
Thermo Scientific™ Nalgene™ PMP Hydrometer Jar
thermo scientific™
N62300500
Protect against breakage. This PMP jar is lighter than glass and offers superior chemical resistance and a broad base, making it a stable addition to the lab or field. Each for $80.17
Case of 6 Each for $481.02