Please login to your online account to display your discounted pricing

Fisherbrand™ Precision Specific Gravity Hydrometers, 150mm Long


Requires 140mL liquid volume for submersion

Manufacturer: fisherbrand™  11556V

 View more versions of this product

Catalog No. 11-556V

  • / Each
 

Description & Specifications

Specifications

Specific Gravity Range 1.84 to 1.92
Increments 0.001
Certifications/Compliance NIST, ASTM E100, ASTM E126
Length (Metric) 165mm
Accessories

Accessories for Catalog Number: 11556V

Item Mfr. No. Item Description Price Per Unit Quantity