Fisher Scientific™ PowerGen Homogenizers Accessories: Saw Tooth Bottom Generator Probes
Saw teeth bottom stainless steel rotor stator probe
$2,127.49 - $4,195.51
Specifications
Bottom Type Flat
Type Flat bottom generator probe
For Use With (Equipment) PowerGen homogenizers manufactured through 2013
Material 316 L Stainless-steel
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Probe Length (Metric) Range Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Probe Length (Metric) Range Price Quantity  
15-340-144 Fisher Scientific Company
FG20195
20mm 195mm 50mL - 2L Each for $2,977.09
15-340-145 Fisher Scientific Company
FG32TT195
32mm 195mm 75mL - 10L Each for $4,195.51
15-340-138 Fisher Scientific Company
FG7195
7mm 195mm 0.25mL - 30mL Each for $2,127.49
15-340-139 Fisher Scientific Company
FG35NA195
35mm 195mm 75mL - 10L Each for $3,735.74
Specifications
Description
  • Material: 316 L Stainless-steel
  • Fits all previous Powergen homogenizers manufactured through 2013
  • For larger sample volumes and sample sizes
  • Resistant to most organic solvents commonly used during homogenization
Description & Specifications
Specifications
Flat
Flat bottom generator probe
PowerGen homogenizers manufactured through 2013
316 L Stainless-steel