Fisher Scientific™ PowerGen™ Cryogenic Homogenizer
Increases sample throughput and helps preserve sample integrity
$1,383.15 - $2,609.65
Specifications
Quantity Each
View More Specs
Includes: Motor unit, tray with six mortars and pestles, Cryogenic Cooler with sample reservoir with mesh cover, absorbent material
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
02-215-500 Fisher Scientific Company
930265
Motor unit, tray with six mortars and pestles, cryogenic cooler with sample reservoir with mesh cover, absorbent material Each for $2,609.65
02-215-502 Fisher Scientific Company
930268
Tray with 6 replacement mortars and 6 pestles Each for $1,383.15
Specifications
Description
Combines the ease and speed of a handheld homogenizer and a liquid nitrogen-cooled sample to homogenize your sample faster and more efficiently.
  • Cordless, compact and versatile
  • Unit's speed-decelerate feature prevents sample splattering and generation of airborne particles
  • Mortar and pestle are constructed of a durable porcelain/zirconium composite
  • Rugged pestle has a stainless-steel shaft with a hex bit for quick connection to the motor
  • Mortar and pestles can be decontaminated between samples by autoclaving, dry heat, or by exposing to a commercial decontaminating solution
  • Cryogenic cooler is valuable for chilling, processing and transporting samples
Order Info
Accessories include tray with six mortars and pestles, and replacement pestles (set of 6)
Description & Specifications
Specifications
Each