Fisherbrand™ PowerGen™ Cryogenic Homogenizer
Increases sample throughput and helps preserve sample integrity.
Manufacturer:  Fisherbrand™ 930268
 View more versions of this product
Catalog No. 02-215-502
Includes: Motor unit, tray with six mortars and pestles, Cryogenic Cooler with sample reservoir with mesh cover, absorbent material
Description
Description
Combines the ease and speed of a handheld homogenizer and a liquid nitrogen-cooled sample to homogenize your sample faster and more efficiently.
  • Cordless, compact and versatile
  • Unit's speed-decelerate feature prevents sample splattering and generation of airborne particles
  • Mortar and pestle are constructed of a durable porcelain/zirconium composite
  • Rugged pestle has a stainless-steel shaft with a hex bit for quick connection to the motor
  • Mortar and pestles can be decontaminated between samples by autoclaving, dry heat, or by exposing to a commercial decontaminating solution
  • Cryogenic cooler is valuable for chilling, processing and transporting samples
Order Info
Accessories include tray with six mortars and pestles, and replacement pestles (set of 6)
Specifications
Specifications
Each
Cryogenic homogenizer
Tray with 6 replacement mortars and 6 pestles
Cryogenic Homogenizing Kit
Documents