Fisher Scientific™ Polypropylene Controlled Heating Mantles
Features a durable, chemically resistant polypropylene outer housing. Fisher Scientific™ Polypropylene Controlled Heating Mantles minimize damage from spills and allow for maximum heat transfer with minimal risk of flask breakage.
Manufacturer:  Fisher Scientific™ EW3600200
 View more versions of this product
Catalog No. 11474111
Specifications Description & Specifications
Specifications
842°F
115V 50/60Hz, 70w
70w
50mL
50mL (1.6 oz.)
Polypropylene housing
450°C
6.8 in.
EM0050/CEX1
17.5cm
1.6 oz.
115V