Please login to your online account to display your discounted pricing
Fisher Scientific™ Paddle Blenders
Unique three-paddle design for homogenizing and mixing samples for microbiological testing
Manufacturer: fisher scientific™  BKL321500F
 View more versions of this product
Catalog No. 11-452-120
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
Capacity 80-400mL
Speed 75-300rpm
Power Consumption 115V/1.25 A