missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ MultiTube Vortexers: Centrifuge Tube Racks
Centrifuge tube racks for Fisherbrand™ MultiTube Vortexers.
Supplier: Fisherbrand™ 80862906
Description
Centrifuge tube racks for Fisherbrand™ MultiTube Vortexers.
For use with Fisher Scientific MultiTube Vortexers
Color-coded foam racks for centrifuge tubes
Specifications
50 mL | |
Centrifuge Tube Rack | |
Multitube vortexer |
50 mL | |
Red | |
17 |