Please login to your online account to display your discounted pricing
Fisher Scientific™ Multi Extraction Mantles: 6 Recess Model, Heating and Stirring
Vented case's unique airflow ensures the unit remains "cool-to-the-touch"
$8,050.19 - $8,050.19
Products
Catalog Number Mfr. No. Capacity Dimensions (L x W x H) Electrical Requirements Model Price Quantity    
09-682-133
fisher scientific™
EMEA61000/CEX1
Each for $8,050.19
Description & Specifications
  • Individual built-in solid state electronic controls enable easy regulation of each heater, while removing sparking associated with mechanical switching
  • Three 12.7mm (0.5 in.) diameter support rods are included
  • Top cover is polypropylene and external surfaces are powder coated and resistant to most chemical solutions
  • Coiled heating element is suspended within a thermal insulating cartridge to provide maximum heat transfer and support, while removing sparking associated with mechanical switching
  • Individual heater cartridges are replaceable
  • Earth (ground) screen encloses the heaters for added safety
  • Neon lights indicate when power is on and supplied to heaters and, if applicable, supplied to stirrers
  • Stirring speed range from 50 to 1000rpm
  • One stirring control operates each set of recesses
  • Stirring module allows the solution to be stirred and heated simultaneously
CE