Fisher Scientific™ Multi Extraction Mantles - 6 Recess Model, Heating Only
Vented case's unique airflow ensures the unit remains “cool-to-the-touch”
$4,311.08 - $4,311.08
Specifications
Design Polypropylene housing
Temperature (Metric) Max 450°C
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity Dimensions (L x W x H) Electrical Requirements Model Price Quantity  
Catalog Number Mfr. No. Capacity Dimensions (L x W x H) Electrical Requirements Model Price Quantity  
09-682-11 Fisher Scientific™
EME60250/CEBX1
250mL (8.4 oz.) 115V 50/60Hz, 900w EME60250/CEBX1 Each for $4,311.08
09-682-141 Fisher Scientific™
EME60100CEBX1
100mL (3.3 oz.) 115V 50/60Hz, 420w EME60100/CEBX1 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Description
  • Individual built-in solid state electronic controls enable easy regulation of each heater, while removing sparking associated with mechanical switching
  • Three 12.7mm (0.5 in.) diameter support rods are included
  • Top cover is polypropylene and external surfaces are powder coated and resistant to most chemical solutions
  • Coiled heating element is suspended within a thermal insulating cartridge to provide maximum heat transfer and support, while removing sparking associated with mechanical switching
  • Individual heater cartridges are replaceable
  • Earth (ground) screen encloses the heaters for added safety
  • Neon lights indicate when power is on and supplied to heaters and, if applicable, supplied to stirrers
Certifications
CE
Description & Specifications
Specifications
Polypropylene housing
450°C