Fisher Scientific™ Multi Extraction Mantles - 3 Recess Model, Heating Only
Vented case's unique airflow ensures the unit remains “cool-to-the-touch”
$2,771.63 - $3,169.25
Specifications
Design Polypropylene housing
Temperature (Metric) Max 450°C
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity Dimensions (L x W x H) Electrical Requirements Model Price Quantity  
Catalog Number Mfr. No. Capacity Dimensions (L x W x H) Electrical Requirements Model Price Quantity  
09-682 Fisher Scientific™
EME30100/CEBX1
100mL (3.38 oz.) 230V 50/60Hz, 180w (UK and EU plug) EME30100/CEBX1 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
09-682-2 Fisher Scientific™
EME30500/CEBX1
500mL (16.9 oz.) 115V 50/60Hz, 600w EME30500/CEBX1 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
09-682-3 Fisher Scientific™
EME31000/CEBX1
1000mL (33.8 oz.) 115V 50/60Hz, 900w EME3 1000/CEBX1 Each for $3,169.25
09-682-1 Fisher Scientific™
EME30250/CEBX1
250mL (8.4 oz.) 115V 50/60Hz, 450w EME30250/CEBX1 Each for $2,771.63
Specifications
Description
  • Individual built-in solid state electronic controls enable easy regulation of each heater, while removing sparking associated with mechanical switching
  • Three 12.7mm (0.5 in.) diameter support rods are included
  • Top cover is polypropylene and external surfaces are powder coated and resistant to most chemical solutions
  • Coiled heating element is suspended within a thermal insulating cartridge to provide maximum heat transfer and support, while removing sparking associated with mechanical switching
  • Individual heater cartridges are replaceable
  • Earth (ground) screen encloses the heaters for added safety
  • Neon lights indicate when power is on and supplied to heaters and, if applicable, supplied to stirrers
Certifications
CE
Warranty
One year parts and labor
Description & Specifications
Specifications
Polypropylene housing
450°C