Fisherbrand™ Moticam™ Digital Cameras
Digital microscope camera with Motic software to create still or moving images on computer
Manufacturer:  Fisherbrand™ MOTICAM2*
 View more versions of this product
Catalog No. 15-000-105
Description
Description
  • Camera with Motic Images Plus software enables to view, capture, enhance, label, measure, print and store images with one program
  • Available with detachable USB cable
  • Generates live, digital output
  • Operating system compatibility: Microsoft Windows XP/Vista/7; Apple Mac OS
  • Operating temperature: -30° to +70°C non condensing
Specifications
Specifications
One year
3.2 x 3.2μm
CMOS
Moticam 2 Digital Microscope Camera
1600 x 1200 (2MP) at 5 fps
Documents