Fisherbrand™ Moticam™ Digital Cameras
Digital microscope camera with Motic software to create still or moving images on computer
Manufacturer:  Fisherbrand™ DMOTICAM10+
 View more versions of this product
Catalog No. 15-000-108
Description
Description
  • Camera with Motic Images Plus software enables to view, capture, enhance, label, measure, print and store images with one program
  • Available with detachable USB cable
  • Generates live, digital output
  • Operating system compatibility: Microsoft Windows XP/Vista/7; Apple Mac OS
  • Operating temperature: -30° to +70°C non condensing
Specifications
Specifications
One year
1.67 x 1.67μm
CMOS
Moticam 10 Digital Microscope Camera
3664 x 2748 (10MP) at 2.2 fps
Documents