Fisher Scientific™ Model 705 Sonic Dismembrator
Ultrahigh power with advanced programming for DNA shearing/ChIP and all liquid processing applications
Manufacturer:  Fisher Scientific™ FB705220
 View more versions of this product
Catalog No. FB705220
Specifications
Description
The Fisher Scientific™ Model 705 Sonic Dismembrator is a full-featured, programmable ultrasonic processor with an easy to use touchscreen control and overload protection. Ultrahigh power with advanced programming for DNA shearing/ChIP and all liquid processing applications.
The additional power and full range of accessories accommodate the widest range of sample volumes and applications. 96 tip horn and Cup Horn accessories offer time saving, high throughput solutions to repetitive, individual sample processing.
  • Processes 0.2 to 1000mL
  • 700 watts
  • Programmable from 1 sec. to 99 hr.
  • Save up to 10 different programs
  • Pulse mode (1 second to 24 hours)
  • Temperature monitoring (requires optional temperature probe)
Applications:
Cell disruption, homogenization, ChIP/DNA shearing, nanoparticle dispersion, protein extraction, sonochemistry, soil extraction, mixing
Description & Specifications
Specifications
230V
700w
50/60Hz
230V 50/60Hz
Generator, converter, 0.5 in. dia. probe, power cable, wrench set
Ideal for cell disruption, homogenization, chip/DNA shearing, nano particle dispersion, protein extraction, sonochemistry, soil extraction, mixing
0.2 to 100mL
1 sec. to 99 hrs.