missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Mini-Tube Rotator Accessories

Interchangeable rotisserie options for all common laboratory tubes (1.5/2.0, 5/7, 15 and 50mL tubes)

Manufacturer:  Fisherbrand™ BKL260753

 View more versions of this product

Catalog No. 05-450-130



Description

Description

 Fisherbrand mini-tube rotator accessory carousels are available in tube capacities 36 of 1.5 to 2mL, 10 of  10/15mL and 16 of  5/7mL, and 6 of 50mL.

  • Ideal for mixing laboratory tubes
  • Cold room and Incubator compatible
  • Adjustable 0 to 90 degrees
  • Durable material
  • 6 of 50mL tubes
Specifications

Specifications

6 tubes
50mL
Fisherbrand mini-tube rotator
6
Carousel
 Mixing
 1EA
Documents