missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Microplate Heaters
Designed for 96-well plate based cell-based and biochemical assays plus 3D cell culture in laminar flow hoods
Manufacturer: Fisherbrand™ 0168R05
Description
The microplate heater maintains consistent temperature across its surface, ensuring cells in outer wells of microplate and inner wells are incubated at same temperature.
- Robust construction
- Achieve higher sensitivity and more rapid results with Thermo Scientific Pierce BCA protein assay kit
- Two button temperature control (ambient +5°C to 65°C (ambient +9°F to 149°F))
- Lightweight with small footprint only slightly bigger than standard microplate
- Clear lid to maintain stable temperature during use in laminar flow hoods and for visibility
- Smooth external surfaces for easy spraying and cleaning
- Max. temperature set point: 65°C (149°F)
- Min. temperature set point: 20°C (68°F)
- Accuracy: ±1°C (±2°F)
- Dimensions: 164 x 147 x135mm (6.4 x 5.7 x 5.3 in.)
- Weight: 1.32kg (2.91 lb.)
Applications:
- Incubating biochemical assays
- Incubating cell based assays
- Trypsinising cells
Specifications
US | |
12 Months | |
1.32 kg |