Please login to your online account to display your discounted pricing
Fisher Scientific™ Micromaster™ Microscope Objectives
Interchangeable DIN objectives for Fisher Scientific Micromaster Series Microscopes
Manufacturer: fisher scientific™  MPOS100
 View more versions of this product
Catalog No. 12-561-100A
  • / Each
Description & Specifications
Specifications
For Use With (Equipment) Micromaster™ infinity optics microscope
Immersion Type Oil
Numerical Aperture 1.25
Product Type achromatic len
Magnification Power 100X