Please login to your online account to display your discounted pricing
Fisher Scientific™ Micromaster™ Microscope Objectives
Interchangeable DIN objectives for Fisher Scientific Micromaster Series Microscopes
Manufacturer: fisher scientific™  MP-OS040
 View more versions of this product
Catalog No. 12-561-40A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
For Use With (Equipment) Micromaster™ infinity optics microscope
Product Type achromatic len
Numerical Aperture 0.62
Magnification Power 40X