Please login to your online account to display your discounted pricing
Fisher Scientific™ Micromaster™ Microscope Objectives
Interchangeable DIN objectives for Fisher Scientific Micromaster Series Microscopes
Manufacturer: fisher scientific™  MPOP025
 View more versions of this product
Catalog No. 12-561-25P
  • / Each
Description & Specifications
Specifications
Product Type Planachromatic lens
Numerical Aperture 0.40
Magnification Power 25X
For Use With (Equipment) Micromaster™ infinity optics microscope