Please login to your online account to display your discounted pricing
Micromaster™ Microscope Eyepiece Reticles
Adapt microscope eyepiece for counting, measuring or positioning
Manufacturer: evos  MP-ERG-2
 View all options for this product
Catalog No. 12-561-RG2
 Please call Customer Service at 1-800-234-7437 or send an email to for assistance.
Description & Specifications
For Use With (Equipment) Eyepiece for Micromaster™ microscope
Product Type Grid reticule
  • Reticles mount inside 21mm I.D.
  • Micromaster I, II or III eyepieces, or 24mm Micromaster infinity eyepieces
  • Monocular Micromaster I, II or III models require focusable eyepiece 12-561-FEP