Fisherbrand™ Maxima C Plus Gasket Sets
Manufacturer:  Fisherbrand™ 0125747
 View more versions of this product
Catalog No. 01-257-47
Description
Description
Contain all the Viton™ O-rings, shaft seals, and gaskets needed to rebuild the pump.
†For M4C, M6C or M8C vacuum pumps designed before September 1999 (S/N before FKK090000).
Specifications
Specifications
Gasket Set
Model M4C, M6C and M4C vacuum pump
Viton™ O-rings, shaft seals and gaskets
Documents