Fisherbrand™ M Series Mechanical Ultrasonic Cleaning Bath
Outfit your lab with a fisherbrand M Series Ultrasonic Cleaner. This unit is designed with solid construction and excellent functionality for a variety of applications − including cell separation, sample preparation and degassing of liquids.
$770.35 - $3,573.65
Specifications
Material Plastic
Timer 60 min.
View More Specs
Products 5
Catalog Number Mfr. No. Model Capacity Dimensions (D x W x H) Interior Voltage Price Quantity  
Catalog Number Mfr. No. Model Capacity Dimensions (D x W x H) Interior Voltage Price Quantity  
15-337-400 View Documents Fisherbrand™
CPX962116R
M1800 1.9L (0.5 gal.) 15 x 14 x 10cm (6 x 5.5 x 4 in.) 120V Each for $770.35
15-337-408 View Documents Fisherbrand™
CPX962216R
M2800 2.8L (0.75 gal.) 23 x 14 x 10cm (9.5 x 5.5 x 4 in.) 120V Each for $1,089.41
15-337-416 View Documents Fisherbrand™
CPX962316R
M3800 5.7L (1.5 gal.) 29 x 15 x 15cm (11.5 x 6 x 6 in.) 120V Each for $1,536.12
15-337-424 Accessories View Documents Fisherbrand™
CPX962516R
M5800 9.5L (2.5 gal.) 29 x 24 x 15cm (11.5 x 9.5 x 6 in.) 120V Each for $2,060.32
15-337-432 View Documents Fisherbrand™
CPX962816R
M8800 20.8L (5.5 gal.) 49.5 x 29 x 15cm (19.5 x 11.5 x 6 in.) 120V Each for $3,573.65
Description
Description
Outfit your lab with a fisherbrand M Series Ultrasonic Cleaner. This unit is designed with solid construction and excellent functionality for a variety of applications − including cell separation, sample preparation and degassing of liquids.
  • Sweep frequency feature eliminates standing waves and creates consistent cavitation throughout tank
  • Controls located above and behind cleaning tank in easily accessible control tower panel
  • Ultrasonic power tracking to track operating frequency of cleaner; always delivering the same power
  • Features 40kHz rugged industrial transducers
  • Chemical-resistant body made of plastic
  • Convenient built-in pour drains present on 0.5 gal. (1.9L) and 0.75 gal. (2.8L) models
  • Tank drains with valves incorporated on 1.5 gal (5.7L) and larger models
  • Includes 60-minute mechanical timer and continuous operation
  • Tank cover included
Order Info
Fisher offers a line of accessories including perforated trays, solid trays, beaker covers and support racks for your application needs.
Specifications
Specifications
Plastic
60 min.