missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Innovating Science™ Acid Spill Cleanup Kit
Contain, remove and tag spills fast.
Manufacturer: Fisher Science Education IS5003
Alert:
WARNING: This product can expose you to chemicals including Respirable crystalline silica, which is known to the State of California to cause cancer.
Description
- One instruction sheet/SDS
- Two gloves
- Two 9 x 16 in. (22.9 x 40.6 cm) polybags
- Two twist ties
- Two blank shipping tags
- One dust pan and brush
- One bag vermiculite absorbent
- One bottle neutralizing mixture (Calcium Hydroxide, Sodium Carbonate, Calcium Carbonate and Litmus Powder)
Specifications
White | |
1 L | |
For labs | |
1 Ea. |
0.26 gal. | |
Acid Spill Kit | |
1 instruction sheet/MSDS, 2 vinyl exam gloves, two 9 x 16 in. polybags, 2 twist ties, 2 blank shipping tags, 1 dust pan and brush, 1 bag vermiculite absorbent, 1 bag neutralizing mixture |
WARNING: This product can expose you to chemicals including Respirable crystalline silica, which is known to the State of California to cause cancer. For more information go to http://www.P65Warnings.ca.gov.