Please login to your online account to display your discounted pricing

Fisher Scientific™ Lab-Sized Spill Cleanup Kits

Contain, remove and tag spills fast.

$67.50 - $71.55

Specifications

Capacity 0.26 gal., 1L
For Use With (Application) For labs
View More Specs
Includes:  Acid Spill Cleanup
One instruction sheet/SDS; two gloves; two 9 × 16 in. (22.9 × 40.6cm) polybags; two twist ties; two blank shipping tags; one dust pan and brush; one bag vermiculite absorbent; one bottle neutralizing mixture (Calcium Hydroxide, Sodium Carbonate, Calcium Carbonate and Litmus Powder).
Caustic Spill Cleanup
One instruction sheet/SDS; two gloves; two 9 × 16 in. (22.9 × 40.6cm) polybags; two twist ties; two blank shipping tags; one dust pan and brush; one bag vermiculite absorbent; one bottle neutralizing mixture (Citric Acid Anhydrous and Litmus Powder).
Solvent Spill Kit
One instruction sheet/SDS; two gloves; two 9 × 16 in. (22.9 × 40.6cm) polybags; two twist ties; two blank shipping tags; one dust pan and brush; one bag diatomaceous earth.
Products
Catalog Number Mfr. No. Includes Product Type Price Quantity    

S80201A

 
fisher scientific™
SS9181
1 instruction sheet/MSDS, 2 vinyl exam gloves, two 9 x 16 in. polybags, 2 twist ties, 2 blank shipping tags, 1 dust pan and brush, 1 bag vermiculite absorbent, 1 bag neutralizing mixture Acid Spill Kit Each for $71.55

S80201B

 
fisher scientific™
SS0917KIT
1 instruction sheet/MSDS, 2 vinyl exam gloves, two 9 x 16 in. polybags, 2 twist ties, 2 blank shipping tags, 1 dust pan and brush, 1 bag vermiculite absorbent, 1 bag neutralizing mixture Caustic Spill Kit Each for $71.55

S80201C

 
fisher scientific™
SS9161
1 instruction sheet/MSDS, 2 vinyl exam gloves, two 9 x 16 in. polybags, 2 twist ties, 2 blank shipping tags, 1 dust pan and brush, 1 bag diatomaceous earth Solvent Spill Kit Each for $67.50
Description & Specifications

Specifications

Capacity 0.26 gal., 1L
For Use With (Application) For labs

Contain enough absorbent and neutralizing agent to safely clean up a 0.26 gal. (1L) spill.