missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Key Chain Laser Pointers

For slide presentations, microscope projections, field studies, wall charts and lectures

Manufacturer:  Fisherbrand™ CON3141

 View more versions of this product

Catalog No. 11-994-2



Description

Description

Key Chain Micro Laser Pointer

  • Half the size of pocket laser
  • Features output power of 4mW and range of 300 ft. (91m)
  • Brilliant red beam with 3.5mm diameter can be seen under normal lighting conditions
  • 2.37L × 0.56 in. dia. (6.0 x 1.4cm) body can be carried securely on keychain
  • Switch is momentary ON/OFF
Laser Key Chain
  • Brilliant red beam works equally well in daylight and dark
  • Easy to carry with attached key chain
  • Highly visible 650nm wavelength
  • Diameter, 3.5mm
  • Output, 4 mW Range, 400 ft.
  • Dimensions, 2.5 × 1 × 0.25 in. (6.35 × 2.54 × 0.64cm)
  • Weight, 0.5 oz. (14.2g)

Specifications

Specifications

Key Chain Laser Pointer
Documents