Please login to your online account to display your discounted pricing

Fisher Scientific™ Key Chain Laser Pointers

For slide presentations, microscope projections, field studies, wall charts and lectures

$115.77 - $115.77

Products
Catalog Number Mfr. No. Price Quantity    

11-994-2

 
fisher scientific™
CON3141
Each for $115.77
Description & Specifications

Key Chain Micro Laser Pointer

  • Half the size of pocket laser
  • Features output power of 4mW and range of 300 ft. (91m)
  • Brilliant red beam with 3.5mm diameter can be seen under normal lighting conditions
  • 2.37L × 0.56 in. dia. (6.0 x 1.4cm) body can be carried securely on keychain
  • Switch is momentary ON/OFF
Laser Key Chain
  • Brilliant red beam works equally well in daylight and dark
  • Easy to carry with attached key chain
  • Highly visible 650nm wavelength
  • Diameter, 3.5mm
  • Output, 4 mW Range, 400 ft.
  • Dimensions, 2.5 × 1 × 0.25 in. (6.35 × 2.54 × 0.64cm)
  • Weight, 0.5 oz. (14.2g)