Please login to your online account to display your discounted pricing

Fisher Scientific™ Jumbo Refrigerator/Freezer Thermometers

Digital thermometers are perfect for monitoring freezers, water baths, heating blocks, incubators and refrigerators. Fisher Scientific Jumbo Refrigerator/Freezer Thermometer accurately monitors temperature with enclosed “temperature-buffered” sensor.

$82.35 - $82.35

Specifications

Certifications/Compliance ISO 9001, DNV, ISO 17025, A2LA, NIST
Accuracy ±1°C
Dimensions (L x W x H) 0.87 x 3.8 x 4.38 in. (2.2 x 9.9 x 11.2cm)
Length (English) Cable 120 in.
Length (Metric) Cable 304.8cm
View More Specs
Includes:  Traceable™ Certificate, flip-open stand for lab bench, slot for wall mounting, one AAA alkaline battery and Velcro™ and magnetic strips to attach to any surface
Products
Catalog Number Mfr. No. Includes Length (English) Probe Length (Metric) Probe Price Quantity    

S95894

 
fisher scientific™
4148/EMD
Traceable certificate, flip open stand for lab bench, slot for wall mounting, one AAA Alkaline battery and Velcro™ and magnetic strips to attach to any surface, with naked sensor 1.5 in. 3.8cm Each for $82.35
Description & Specifications

Specifications

Certifications/Compliance ISO 9001, DNV, ISO 17025, A2LA, NIST
Accuracy ±1°C
Dimensions (L x W x H) 0.87 x 3.8 x 4.38 in. (2.2 x 9.9 x 11.2cm)
Length (English) Cable 120 in.
Length (Metric) Cable 304.8cm
Resolution 0.1°C
Weight (English) 4 oz.
Weight (Metric) 113g

Bottle insulates sealed sensor from rapid temperature changes when refrigerator door is opened. Thermometer fulfills all CDC thermometer and vaccine thermometer requirements.

 

  • Easy-view, jumbo 1.5 in. (3.8cm) digits are able to be read from 30 ft. (9.1m)
  • Sensor and 10 ft. (3m) cable perform accurately even when both are under water
  • High-impact, chemical-resistant ABS plastic case
  • Available with sealed probe or naked sensor

Record Maximum/Minimum Readings
  • Triple display simultaneously shows maximum, minimum and current probe temperatures plus room temperature
  • Min./max. monitors high/low readings overnight, on weekends or for any time period — a significant advantage over “current reading-only” glass thermometers
  • Range is -58° to +158°F and -50° to +70°C with a resolution of 0° and accuracy of ±1°C
Superior Design for Safe Operation
  • Alarm feature provides alert when temperature rises above or falls below a set point
  • Alarm is programmable in 1° increments
  • 10 ft. micro-cable permits refrigerator doors to close on it
  • Solid-state probes eliminate mercury contamination in refrigerators
  • Sealed probe is housed in a miniature bottle filled with nontoxic glycol; solution is GRAS (generally recognized as safe) by the FDA (Food and Drug Administration)
  • Eliminates concerns about incidental contact with food or drinking water

Case: 3.88 × 4.38 × 0.88 in. (9.9 × 11.2 × 2.2cm). Sealed bottle probe measures 1 × 2.5 in. (2.5 × 6.4cm). Naked sensor is 0.19 dia. × 0.75 in. (4.8 × 19mm) long.

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.