Please login to your online account to display your discounted pricing
Fisher Scientific™ Isotemp™ Microbiological Incubators
Precise, repeatable, thriving incubation
$3,039.92 - $4,654.20
Specifications
Voltage 120V
Hertz 60Hz
Plug Type NEMA 5-15 US
Max. Shelf Load 25 lb. (55kg)
View More Specs
Products
Catalog Number Mfr. No. Volume (Metric) Convection Technology Price Quantity    
151030513
  Accessories
fisher scientific™
51030513
75L Gravity Each for $3,039.92
151030514
fisher scientific™
51030514
117L Gravity Each for $3,774.46
151030515
  Accessories
fisher scientific™
51030515
194L Gravity Each for $4,275.48
151030517
fisher scientific™
51030517
104L Forced Air Each for $4,363.32
151030516
fisher scientific™
51030516
66L Forced Air Each for $3,839.14
151030518
fisher scientific™
51030518
178L Forced Air Each for $4,654.20
Description & Specifications
Specifications
Voltage 120V
Hertz 60Hz
Plug Type NEMA 5-15 US
Max. Shelf Load 25 lb. (55kg)
  • Outstanding temperature uniformity and stability to keep samples at desired condition    
  • Wide temperature range: ambient +5°/10° to 75°C    
  • Intuitive and easy-to-use microprocessor controller    
  • Automatic over temperature alarm—no operator settings required    
  • Low air exchange provides minimal drying out of samples—ideal for long experiments    
  • On/off timer    
  • Corrosion resistant stainless steel interior with smooth corners for easy cleaning†    
  • Easy-to-clean rounded interior edges, designed to reduce contamination risks    
  • Flexible shelving with no tools required to set up, adjust or remove    
  • Small external dimensions per chamber size reduce required bench space
Construction Features:
  • Stainless-steel exterior provides a clean professional look with surface that avoids fingerprint smudges
  • Readily stackable same-size units without the need for tools or kits
  • Access port with silicone plug for independent data logging or for using small equipment inside the unit
  • Flexible shelving system with different positions to optimize chamber space
  • Large easy-to-view fluorescent display with intuitive microprocessor controller
  • Internal glass door for undisturbed sample viewing
cCSAus certified
2 year parts and labor warranty
† Gravity convection models AISI 430 / 1.4016 and mechincal convection (forced air) models AISI 304 / 1.4301
‡ Depth of handle / display not included in depth (65mm / 2.6 in.); adjustable feet not included in height (35mm / 1.4 in.)—required distance to rear wall: 80mm / 3.1 in.