Fisher Scientific™ Isotemp™ Large-Capacity Refrigerated Incubator Shelves
Add up to 33% extra capacity
$256.22 - $256.22
Specifications
Product Type Shelf
Material Steel Wire
Width (English) 31.43 in.
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Holds Length (English) Price Quantity  
Catalog Number Mfr. No. Holds Length (English) Price Quantity  
11-679-10L Fisher Scientific™
1167910L
Up to 88 BOD bottles 21.56 in. Each for $256.22
Specifications
Description
  • Made of coated steel wire
  • Large shelf fits lower half of unit, holds up to 88 BOD bottles; measures 219/16L × 317/16 in.W (54.8 × 79.9cm)
  • Small shelf fits upper half of unit, holds up to 77 BOD bottles; measures 195/16L × 317/16 in.W (49 × 79.9cm)
Description & Specifications
Specifications
Shelf
Steel Wire
31.43 in.