missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisher Scientific™ Isotemp™ Large-Capacity Refrigerated Incubator Chart Recorder Paper

For 11-679-304RN.

$94.23 - $94.23

Specifications

Diameter (English) Base 6 in.
Diameter (Metric) Base 15cm
Product Type Chart Recorder Paper
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Temperature Range (English) Temperature Range (Metric) Price Quantity  
Catalog Number Mfr. No. Temperature Range (English) Temperature Range (Metric) Price Quantity  
15-074-79 Fisher Scientific™
C651
-50 to 50°F -50 to 50°C Pack of 60 Each for $94.23
15-074-80 Fisher Scientific™
C653
0 to 50°F 0 to 50°C Pack of 60 Each for $94.23
Specifications

Description

  • Circular, seven-day, 6 in. diameter (15cm)
  • Range is user determinable
  • Comes in package of 60 sheets
Description & Specifications

Specifications

6 in.
15cm
Chart Recorder Paper