Fisher Scientific™ Isotemp™ Hotplate
Experience exceptional safety and temperature performance for your routine protocols.
$630.22 - $1,065.28
Products ${productFamilyLength}
Catalog Number Mfr. No. Electrical Requirements Plug Type Surface Area (English) Heating Top Plate Material Price Quantity  
Catalog Number Mfr. No. Electrical Requirements Plug Type Surface Area (English) Heating Top Plate Material Price Quantity  
HP88850200 Fisher Scientific™
HP88850200
100-120V 50/60Hz US style 10.25 x 10.25 in. Ceramic Each for $1,065.28
HP88854200 Fisher Scientific™
HP88854200
100-120V 50/60Hz US style 4.25 x 4.25 in. Ceramic Each for $630.22
HP88857200 Fisher Scientific™
HP88857200
100-120V 50/60Hz US style 7.25 x 7.25 in. Ceramic Each for $716.29
HP88857204 Fisher Scientific™
HP88857204
100-120V 50/60Hz US style 7.25 x 7.25 in. Aluminum Each for $674.00
Specifications
Description
Experience exceptional safety and temperature performance for your routine protocols. Available in three sizes, providing flexibility from microscale chemistries to production operations.
Advanced technology
  • Strong magnetic coupling
  • Temperature adjustable in 1 degree increments
  • HOT TOP warning system designed to protect from accidental burns with prominent display when heating surface is above 50°C (122°F)
  • Automatic over-temperature protection
Flexible, robust design
  • Top plate choices in ceramic or aluminum
  • Raised display design protects electronics from spills
  • Optional temperature probe and splash guard protection shield
  • Variable sizes for different sample volumes
Certifications
cCSAus (100-120V models), CE (230V models)
Compliance
RoHS compliant
Warranty
Three years