Fisher Scientific™ Isotemp™ Heating Blocks
For Isotemp digital models 125DQ, 145DQ and analog dry-bath incubators
Manufacturer:  Fisher Scientific™ 2075FSQ
 View more versions of this product
Catalog No. 11715319Q
Specifications Description & Specifications
Specifications
Isotemp 125DQ, 145DQ and economical dry-bath incubator
2 in.
2.9 in.
Aluminum
9.5cm
1.96 in.
For 6 x 25mm tube
Heating Block
2.9 in.
2.9 x 3.75 x 2 in. (7.4 x 9.5 x 5.1cm)
7.4 x 9.5 x 5cm
5cm
5.1cm
7.4cm
3.75 in.
9.5cm
50mm
2 in.
51mm
74mm
2.9 x 3.75 x 2 in.
7.4cm