Please login to your online account to display your discounted pricing
Fisher Scientific™ Isotemp™ General-Purpose Series Lab Freezers, 24 cu. ft.
Reliable cold storge for general-purpose applications
Specifications
Certifications/Compliance UL
Defrost Automatic
Hertz 60Hz
Insulation Polyurethane Foam
Refrigerant R-404A
View More Specs
Products
Catalog Number Mfr. No. Includes Price Quantity    
11670202
fisher scientific™
MF25SSSAEEFS
Alarms CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
11670203
fisher scientific™
MF25SSSAREFS
Alarms and Chart Recorder CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
Certifications/Compliance UL
Defrost Automatic
Hertz 60Hz
Insulation Polyurethane Foam
Refrigerant R-404A
Temperature Sensor Digital
Isotemp General- Purpose Series freezers offer the dependability and safety required for your daily lab applications.
  • 24 cu. ft. capacity
  • Freezer cabinet range: -12° to -30°C (factory setpoint -30°C)
  • Lockable doors are manufactured in durable, stainless steel
  • Stainless-steel interior
  • Standard access port
International models are available; contact your Fisher Scientific Customer Service Representative for details.
Service agreement available.
UL Listed
13 months, parts and labor
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.