missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisher Scientific™ Isotemp™ Flammable-Materials Storage Refrigerators: Basic Model

For reliable, safe storage of volatile materials, pharmaceuticals, clinical reagents, and biological samples

$4,023.93 - $6,836.10

Specifications

Defrost Manual Defrost
Insulation Polyurethane Foam
Refrigerant R134a, 100% CFC Free
Temperature Range (Metric) 1° to 12°C
Certifications/Compliance UL listed meets NFPA 45 and 70
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Electrical Requirements Capacity Door Count Shelves Price Quantity  
Catalog Number Mfr. No. Electrical Requirements Capacity Door Count Shelves Price Quantity  
05FREEFSA Fisher Scientific™
05FREEFSA
115 V/60 Hz 5.5 Cubic Feet/155.7 Liters 1(1 compartment) 3 Adjustable Shelves, Storage On Bottom Of Unit Each for $4,023.93
50FREEFSA Fisher Scientific™
50FREEFSA
115 V/60 Hz 50 Cubic Feet/1415.8 Liters 1(1 compartment) 8 Adjustable Shelves N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
20FREEFSA Fisher Scientific™
20FREEFSA**
115 V/60 Hz 20.9 Cubic Feet/591.82 Liters 1(1 compartment) 4 Fixed Shelves, 6 Fixed Door Shelves Each for $6,836.10
Specifications

Description

  • Adjustable temperatures from 1° to 12°C, depending on model
  • 50 cu. ft. (1420L) model features digital temperature display in 0.1° increments, °C or °F, and keyed power switches
  • Manual defrost
Safety Features
  • Orange warning label on front
  • Sparkproof interiors separate contents from compressor and motor
  • Compressors have enclosed overloads, relays, and thermostats that prevent sparks from escaping during door opening and closing
  • Static hot-wall condensers and evaporators are free of electric fans
  • Magnetic door gaskets provide airtight seals that insulate and prevent release of hazardous fumes
  • Top-mounted hydraulic thermostat and door stop
Construction
  • White, baked-on polyester enamel exteriors CFC-free polyurethane foam insulation and refrigerant
  • Solid door(s)

Service Plan

Service agreement available.

Certifications

UL listed meets NFPA 45 and 70

Compliance

NFPA Code 99 (30 and 50 cu.ft. models only)

Warranty

13 months, parts and labor

† Allow 3 in. (7.6cm) clearance top, back, and sides.
†† With legs. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

Manual Defrost
R134a, 100% CFC Free
UL listed meets NFPA 45 and 70
Polyurethane Foam
1° to 12°C