Please login to your online account to display your discounted pricing
Fisher Scientific™ Isotemp™ Explosion-Proof Refrigerator
Ideal for storing volatile materials in areas with explosive atmospheres or in hazardous locations
$6,022.33 - $6,022.33
Catalog Number Mfr. No. Electrical Requirements Capacity Price Quantity    
fisher scientific™
120V/60Hz 5.5 cu. ft. / 155.7L Each for $6,022.33
Description & Specifications
Prevent the triggering of an explosion inside or outside the unit. Fisher Scientific™ Isotemp™ Explosion-Proof Refrigerator is suitable for use in Class I, Division I Group C and D hazardous environments. Thermostat and compressor components are isolated within a vapor-proof enclosure, and electrical connections are hardwired to prevent sparking and accidental disconnection.
  • Hydraulic thermostat
  • Manual defrost
  • Magnetic vinyl gaskets for positive door seal
  • CFC-free insulation and refrigerant
Not equipped with line cord and plug; must be hardwired into an appropriately rated conduit box.
Service agreement available.
UL listed meets NFPA 45 and 70
13 months, parts and labor