Please login to your online account to display your discounted pricing

Fisher Scientific™ Isotemp™ Advanced Hotplate

Reliable performance, dependable safety and precise control. Available with optimized controls and settings for applications that demand advanced precision.

$1,562.24 - $1,868.28

Specifications

Surface Area (English) Heating 7.25 x 7.25 in.
Surface Area (Metric) Heating 18.4 x 18.4cm
Dimensions (L x W x H) 13 x 8.2 x 3.8 in. (33 x 20.8 x 9.7cm)
Description Hotplate
View More Specs
Products
Catalog Number Mfr. No. Electrical Requirements Plug Type Max. Temperature (Metric) Top Plate Material Price Quantity    

HP88857290

 
fisher scientific™
HP88857290
100-120V 50/60Hz US style 450°C Ceramic Each for $1,562.24

HP88857294

 
fisher scientific™
HP88857294
100-120V 50/60Hz US style 300°C Aluminum Each for $1,868.28
Description & Specifications

Specifications

Surface Area (English) Heating 7.25 x 7.25 in.
Surface Area (Metric) Heating 18.4 x 18.4cm
Dimensions (L x W x H) 13 x 8.2 x 3.8 in. (33 x 20.8 x 9.7cm)
Description Hotplate

Intuitive design
  • Top plate choices in ceramic or aluminum
  • Raised display design protects electronics from spills
  • Ability to set programs (ramp/dwell) as well as save settings
  • Includes stainless-steel PT100 temperature probe, supporting rod and clamp kit
  • Optional PT1000 temperature probe and splash guard protection shield
Advanced technology
  • HOT TOP warning system designed to protect from accidental burns with prominent display when heating surface is above 50°C (122°F)
  • Temperature adjustable in 1°C increments except when using temperature probe then ability to control in 0.1 degree increments below 100°C

cCSAus (100-120V models), CE (230V models)

RoHS compliant

Three years