Please login to your online account to display your discounted pricing

Fisher Scientific™ Isotemp™ Digital and Analog Dry Bath Incubators

Provide controlled dry heat for a wide variety of clinical and general chemistry applications

$941.46 - $1,595.61

Specifications

Material Powder coated Steel
Temperature Range (Metric) 5 to 130°C
Display LED
Electrical Requirements 120V 50/60Hz
Hertz 50/60Hz
View More Specs
Includes:  Digital Models come with temperature probe, line cord and plug. Analog Models include high and low thermostat, power switch, pilot light, line cord and plug.
Requires:  Heating blocks, sold separately, hold regular tubes, microcentrifuge tubes, square cuvets, or microplates.
Products
Catalog Number Mfr. No. Amperage Dimensions (L x W x H) Temperature Uniformity Wattage Price Quantity    

117184Q

  Accessories
fisher scientific™
2053FSQ
1.67A 9.13 x 11.25 x 3.38 in. (23.1 x 28.6 x 8.6cm) ±0.5 at 37°C 200w Each for $941.46

117186Q

  Accessories
fisher scientific™
2054FSQ
2.5A 12.38 x 8.31 x 3.38 in. (31.5 x 21.1 x 8.6cm) ±0.5 at 37°C 300w Each for $1,595.61
Description & Specifications

Specifications

Material Powder coated Steel
Temperature Range (Metric) 5 to 130°C
Display LED
Electrical Requirements 120V 50/60Hz
Hertz 50/60Hz
Voltage 120V
Warranty 1 Year

Choice of 1- to 6-block Analog Models, 2- or 4-block Digital Models.

  • Exceptional temperature control and uniformity
  • Temperature range of 5°C above ambient to 130°C
  • Powder-coated steel body construction provides durability for demanding applications
  • Compact design saves valuable bench space
  • Choice of interchangeable heating blocks

Digital Dry Baths with Microprocessor Controller

  • PID microprocessor control, digital temperature setting
  • Just press up/down arrows to set temperature in 0.1° increments
  • Effectively hold samples at constant, warm temperature
  • 5°C above ambient to 130°C range; ±0.4° uniformity at 37°C; stability, ±0.2°C
  • Bright LED display continuously shows actual temperature in °C
  • Temperature probe in bottom of bath provides improved accuracy and control; free probe included
  • Model 125DQ holds two heating blocks; Model 145DQ holds four heating blocks in 2 × 2 configuration
Analog Dry Baths with Dual-thermostat Controls
  • Sloped control panel makes controls highly visible
  • Dual bimetallic thermostat controls
  • Stability is ±1.5°C, uniformity ±0.5°C at 37°C
  • Use thermometer to read current temperature of blocks, then adjust control to reach desired setpoint
  • Efficient design: operate at substantially lower external temperature than other models

Applications:

Ideal for enzyme reactions, inactivation of sera, Rh studies, blood crossmatching and cholesterol determinations

All models accept 2.91L × 3.75W × 2 in.H (7.4 × 9.5 × 5.1cm) blocks, sold separately.

Digital Models are CSA approved. Analog Models are UL listed.

One year

†International model