Please login to your online account to display your discounted pricing

Fisher Scientific™ Incubating/Cooling Mini Shakers

Microplate ready without needing additional accessories

$6,847.08 - $6,847.08

Specifications

Dimensions (L x W x H) 17.9 x 11 x 10.5 in. (45.5 x 27.9 x 26.7cm)
Humidity Range 80%
Temperature Range (English) 41 to 104°F
Temperature Range (Metric) 5 to 40°C
Timer 1 sec. to 160 Hours
View More Specs
Products
Catalog Number Mfr. No. Amperage Voltage Price Quantity    

02-217-755

 
troemner llc
980FIICMDDUS
2A 120V Each for $6,847.08
Description & Specifications

Specifications

Dimensions (L x W x H) 17.9 x 11 x 10.5 in. (45.5 x 27.9 x 26.7cm)
Humidity Range 80%
Temperature Range (English) 41 to 104°F
Temperature Range (Metric) 5 to 40°C
Timer 1 sec. to 160 Hours
Wattage 200w
Display LEDLED
Includes Polycarbonate lid
Speed 100 to 1200rpm
Wash/Waste Bottle Capacity Two microplates/modular blocks

The instrument provides consistent and uniform shaking action. Microprocessor displays last set point and restarts if power is interrupted. Easy-to-use controls allow users to adjust temperature in 1°C increments. Provides repeatable and accurate results every time. Display is easily visible across lab benches. Can be used for analyses that require stable and controlled temperatures.

  • Features variable speed microprocessor control
  • PID temperature controller for precise control from ambient +5° to +65°C
  • Triple eccentric drive with lubricated ball bearings and maintenance free, brushless DC motor
  • Easy-to-read, independent LED displays for temperature, speed and time
  • Temperature calibration mode to calibrate unit to an external temperature device
  • Overload protection by activation of audible and visual signals
  • Speed ramping feature to slowly increase speed to desired set-point
  • Timer with audible alarm

  • Caution hot indicator symbol
  • Spill-resistant design
  • Polycarbonate lid enables viewing samples without disturbing internal temperature
  • Operating conditions: 5° to 40°C (41° to 104°F), maximum 80% relative humidity, non-condensing

Cell and bacterial cultures, hybridizations and enzyme reactions

Includes 92 in. (234cm) detachable, 3-wire cord and plug (230V units are supplied with Euro type plug) and adapter bracket to hold optional modular blocks

CE marked, TUV listed

2 year limited warranty on parts and labor