Fisherbrand™ Homogenizer Model 1000 Package
For fast and economical dispersion of both small and large amount of products
Supplier:  Fisherbrand™ 61090050
Catalog No. 08-451-672
Description
Description
Homogenizer Model 1000
  • With integrated electronic speed regulation and stabilization
  • Potentiometer allows for easy setting of desired speed
  • Has automatic soft start and integrated thermal overload fuse which protect motor and ensures longevity
  • Available with user friendly coupling that allows easy exchange of generators
  • Production of dispersions and emulsions
  • Dispersion of plant, human or animal tissues, resins and pigments
  • Dissolving solids and production of suspensions
  • Dispersion of APIs in pharmaceutical preparations
  • Formulation of lotions, creams and food products
  • Production of oil /water emulsions
  • For extraction of active substances, QA and QC of certain products
Specifications
Specifications
Homogenizer Package
115 V
Ideal for production of dispersions and emulsions
Model 1000 homogenizer (115 V), stand