Please login to your online account to display your discounted pricing

Fisher Scientific™ Homogenizer Model 1000 Package

For fast and economical dispersion of both small and large amount of products

$2,502.40 - $2,502.40

Specifications

For Use With (Application) Ideal for production of dispersions and emulsions
Type Homogenizer Package
View More Specs
Includes:  Fisher Scientific homogenizer model 1000 (115V or 230V), stand
Products
Catalog Number Mfr. No. Includes Voltage Price Quantity    

08-451-672

 
fisher scientific™
61090050
Model 1000 homogenizer (115V), stand 115V Each for $2,502.40
Description & Specifications

Specifications

For Use With (Application) Ideal for production of dispersions and emulsions
Type Homogenizer Package

Homogenizer Model 1000

  • With integrated electronic speed regulation and stabilization
  • Potentiometer allows for easy setting of desired speed
  • Has automatic soft start and integrated thermal overload fuse which protect motor and ensures longevity
  • Available with user friendly coupling that allows easy exchange of generators

  • Production of dispersions and emulsions
  • Dispersion of plant, human or animal tissues, resins and pigments
  • Dissolving solids and production of suspensions
  • Dispersion of APIs in pharmaceutical preparations
  • Formulation of lotions, creams and food products
  • Production of oil /water emulsions
  • For extraction of active substances, QA and QC of certain products