Bio Plas™ HistoPlas™ Tissue Processing Capsules
The integral lid fits tightly—you never need to worry about losing your tissues
Manufacturer:  Bio Plas™ BIP6010F
 View more versions of this product
Catalog No. 15-182-218
Description
Description
The integral lid fits tightly—you never need to worry about losing your tissues
  • Disposable polypropylene capsules ensure proper fluid exchange in processing and infiltration
  • Open-mesh base and lid allow superior flow and drainage
  • One-piece design is hinged for perfect lid/base alignment
  • With generous labeling surface for quick identification
Specifications
Specifications
White
Tissue capsules
1000/Cs.
Documents