Please login to your online account to display your discounted pricing
Fisher Scientific™ Heating/Cooling Mini Shakers for PCR
Ideal for analyses in a stable, controlled environment and PCR plate ready without the need for any additional accessories
$2,336.76 - $2,336.76
Products
Catalog Number Mfr. No. Price Quantity    
88861039
fisher scientific™
88861039
Each for $2,336.76
Description & Specifications
  • Microprocessor control - The variable speed microprocessor control provides consistent, uniform shaking action. Microprocessor will display last set-point and will restart if power is interrupted.
  • PID temperature controller - Maintains precise temperature control from 10°C below ambient to 65°C. Easy-to-use controls allow users to adjust temperature in 1°C increments.
  • Triple eccentric drive - Permanently lubricated ball bearings and maintenance free, brushless DC motor provide reliable service and continuous duty operation.
  • LED display - Touch pad controls with easy-to-read, independent LED displays for temperature, speed and time allow operator to view all settings at once. Provides repeatable and accurate results every time and is easily visible across lab benches. Timer will display elapsed time or, when programmed to user defined limit, will shut off unit when time reaches zero. Display will show last used settings, even after power has been turned off.
  • Temperature calibration mode - Allows user to calibrate unit to an external temperature device
  • Applications - ELISA, Enzyme Activity Analysis, Enzyme cutting, DNA ligation, PCR