Fisherbrand™ Hands-Free Fold-Up Magnifiers
Allow two-handed examination of objects while yielding a sharp, crisp and distinct image
Manufacturer:  Fisherbrand™ CON3434
 View more versions of this product
Catalog No. 14-648-23
Description
Description
Fisher Scientific™ Hands-Free Fold-Up Magnifiers all fold twice for compact storage. Allow two-handed examination of objects while yielding a sharp, crisp and distinct image.
  • Bottom panel features white grid markings in both inches and millimeters to assist in measuring
  • All-glass optics ensure perfect focus with zero distortion
  • Seven sizes are available, including three featuring brilliant battery-powered illumination
Specifications
Specifications
1.25 in.
For use in quality control, labs, food, electronics, manufacturing and universities
5x
3.17cm
Glass
Hands-Free™ Fold-up Magnifier
Documents